Would you like to log in again?
DEV Tryptophan Broth Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
GM1901-2.5KG GM1901-5KG GM1901-500G
75804-238EA 700.46 USD InStock
75804-238 75804-240 75804-242
DEV Tryptophan Broth Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
DEV tryptophan broth medium can be used for the subcultivation of coliform, differentiation and for indole testing in the bacteriological examination of water.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty