Would you like to log in again?
TAT Broth Base, HiMedia Laboratories
Supplier: HiMedia Laboratories
GM562-500G GM562-5KG GM562-2.5KG
75804-298EA 732.46 USD InStock
75804-298 75804-300 75804-302
TAT Broth Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
TAT Broth base, granulated with added polysorbate 20 is used for sterility testing of highly viscous or gelatinous substances such as salves, ointments and other cosmetic products.
  • Off-white to yellow colored
  • Granular medium
  • Reaction of 2.5% w/v aqueous solution containing 4% v/v polysorbate 20 at 25 °C
Caution: Store below 30 °C in tightly closed container and prepared medium at 2 to 8 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty