Would you like to log in again?
Lactobacillus MRS Agar Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M641I-500G M641I-5KG M641I-2.5KG M641I-5KG M641-500G M641I-500G M641-500G GM641-500G M641I-2.5KG GM641-5KG GM641-2.5KG
89226-118EA 1386.13 USD InStock
CA89226-118 89226-118 CA89226-116 75804-360 CA95022-948 75804-362 CA95022-946 95022-948 95022-946 89226-116 75804-358
Lactobacillus MRS Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Lactobacilli MRS Agar is recommended for the isolation, enumeration and cultivation of Lactobacillus spp.
  • Final pH ( at 25 °C) is 6.5 ±0.2
Certifications: The composition and performance criteria of M641I is according ISO 1995, Draft ISO/DIS 13720.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty