Would you like to log in again?
Brilliant Green Agar Base with 1.2% Agar Dehydrated Culture Media, Granulated, HiMedia Laboratories
Supplier: HiMedia Laboratories
GM016A-5KG GM016A-500G GM016A-2.5KG
75804-490EA 839.07 USD InStock
75804-490 75804-492 75804-494
Brilliant Green Agar Base with 1.2% Agar Dehydrated Culture Media, Granulated, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Recommended as an enrichment medium for the isolation of Salmonellae from feces, urine, and other pathological materials.
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty