Would you like to log in again?
SAMPLER AIR DUO TRIOBAS CONT 200LTS/MI
Supplier: HARDY DIAGNOSTICS
Trio.Bas* Duo Air sampler 200lts/min^ Trio.Bas* Duo Air sampler, With Bluetooth, RODAC plate, Contact Plate brackets, Shockproof, IP65 protection from dust and water, Flow rate: 200lts/min
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SAMPLER AIR DUO TRIOBAS CONT 200LTS/MI 75804-720 BAS225 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty