Would you like to log in again?
TRIO.BAS™ SELFTEST Base Station Induction Battery Charger Calibration Check System
Supplier: Hardy Diagnostics
TRIO.BAS™
BAS352 BAS351
75804-760EA 7881.5 USD InStock
75804-760 75804-762
TRIO.BAS™ SELFTEST Base Station Induction Battery Charger Calibration Check System
Samplers Air Samplers
The Base Station Induction Battery Charger with user Self-test is equipped with a system that allows users to check the state of precision of the airflow, regardless of auto-calibration already present in the instrument. This verification is necessary to avoid the possible invalidation of the tests between annual controls for official certification.
  • Calibration bell
  • Connector cable and charger with outlet plug
This base is used in replacement of the base station induction charger when the Self-test system is applied.
Note: this unit does not take the place of official calibration services as outlined by the USP.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty