Would you like to log in again?
SteriKit® Sterile Vials, Stoppers, and Seals, Prince Sterilization Services
Supplier: Prince Sterilization Services
037 023 034 003 004 002 102 035 009 075 005 012 010 022 025 007 008 036 024 067 018 006 001 011
75805-162CS 5748.51 USD InStock
75805-162 75805-172 75805-150 75805-160 75805-170 76614-772 76614-782 76614-770 76614-780 76614-776 75805-158 76614-774 75805-168 75805-156 75805-178 76614-784 75805-166 75805-154 75805-176 76614-768 76614-778 75805-164 75805-152 75805-174
SteriKit® Sterile Vials, Stoppers, and Seals, Prince Sterilization Services
Vials
The SteriKit® consists of ready to use vials, seals, and stoppers.
The vials are type I borosilicate glass and are USP WFI washed, aseptically repackaged in a 316 stainless steel tray, and USP dry heat depyrogenated in an ISO 7 cGMP cleanroom. The stoppers and seals are USP WFI washed, aseptically repackaged, and cGMP moist heat steam sterilized. Following sterilization, all components are assembled, vacuum packed, and heat sealed in a pre-sterilized LDPE bag under ISO 5 conditions.
All components are certified in a cGMP laboratories to be USP 85 pyrogen-free, USP 788 particulate-free and USP 71 sterile. A certificate is available with each order. The SteriKit® is completely customizable, and available with vial sizes ranging from 2 to 100 ml. The SteriKits® are double vacuum packaged under sterile ISO 5 conditions. The disposable packaging is easy to use.
Certifications: Certified per EP 2.9.19-Test 1B and USP 788 for particulates, EP 2.6.14 and USP 85 for bacterial endotoxins, and EP 2.6.1 and USP 71 for sterility testing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty