Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
RT Multi-Position Hotplate Stirrers, IKA Works
Supplier: IKA
0003690601 0003692601 0003691101
75805-610EA 5732.15 USD InStock
75805-610 75805-608 75805-606
RT Multi-Position Hotplate Stirrers, IKA Works
Stirrers Magnetic Hotplate Stirrers, Multi-position
5, 10, or 15-position digital magnetic hotplate stirrers, designed for synchronous heating and stirring
The magnetic coil technology provides noiseless and consistent stirring on all positions. The surface temperature is infinitely adjustable up to 120°C, producing a maximum medium temperature of 70°C (depending on the used vessel). Speed remains constant, even when load changes
Certifications: RoHS compliant.
Ordering information: Unit comes with detachable power cord.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty