Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Direct red 80 kit stain for histology

Supplier: POLYSCIENCES INC-RESEARCH CHEM
Synonyms: CI 35780Sirius RedSirius Red F3BPicro-Sirius Red

24901-250 24901-500
75805-980KT 718.32 USD InStock
75805-980 75805-978
Direct red 80 kit stain for histology
Direct Red 80
This stain binds specifically to collagen fibrils of varying diameter to distinguish collagen type I from type III

This stain helps in quantifying the amount of collagen in a given area of myocardial tissue and is used to analyse the quantitative aspect of liver fibrosis.

Delivery information: Kit contains Phosphomolybdic Acid, Picrosirius Red F3BA Stain, 0.1 N Hydrochloride Acid.


Formula: C₄₅H₂₆N₁₀Na₆O₂₁S₆
MW: 1373.051 g/mol
MDL Number: MFCD00054389
CAS Number: 2610-10-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty