Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ice Borer, Bürkle

Supplier: Bürkle

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

5323-2010 5323-0200 5323-0190
75806-596EA 1205.62 USD InStock
75806-596 76501-442 75806-594
Ice Borer, Bürkle
Samplers Solid Samplers
Sampler for deep-frozen and semi-solid substances products in the food industry.

  • Stainless steel AISI 316L (1.4404) / AISI 316 Ti (1.4571)
  • Sterilizable
  • Complete with case

The sharp sawing crown of the hollow borer rapidly cuts into the sample

It consists of three parts: borer, borer head for use in conventional drills and rod for pressing the cylindrical sample out of the borer head.

A cylindrical sample (cross sectional sample) with a diameter of 1.6 cm and a length of up to 20 cm can be cut out.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty