Would you like to log in again?
Surgipath® Parafree Stainless Steel Base Molds, Leica
Supplier: Leica
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Surgipath®
39LC-700-4 39LC-700-2 39LC-700-5 39LC-700-3
75806-630BX 138.78 USD InStock
75806-630 75806-626 75806-628 75806-624
Surgipath® Parafree Stainless Steel Base Molds, Leica
Moulds Moulds for Histology
Parafree Stainless Steel Base Molds offer higher walls then other base molds in the market, eliminating excess paraffin build-up around the block.
  • Available in 4 sizes
  • Unique design
  • 80% less block scraping
  • Precise corners
The unique design of these base molds produces paraffin blocks that require 80% less block scraping before sectioning. The precise corners of the mold help to facilitate immediate ribboning.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty