Would you like to log in again?
COLUMN PEP-2 SPE 150MG/6ML 80UM 70A PK30
Supplier: Agela Technologies
Cleanert* PEP-2 SPE Cloumn 150 mg^ Cleanert* PEP-2 SPE Cloumn, Membrane: Polyethylene Frit, Reversed-phase, used for trace analysis of food and environmental samples, Particle Size: 80um, Pore Size: 70 armstrong, Volume: 6mL, Size: 150mg
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN PEP-2 SPE 150MG/6ML 80UM 70A PK30 75806-924 PE1506-2 Pack of 30 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty