Would you like to log in again?
COLUMN DURASHELL C18 ANAL 3UM 3.0X100MM
Supplier: Agela Technologies
Column Durashell C18 3UM^ Column Durashell C18, Surface Area: 380 m2/g, Dimension: 3.0X100MM, Features: Wide pH range (1.5-12.0), Minimal silanol activity, Better efficiency than many other high pH compatible columns, High loading capacity for basic compounds for preparative apps, Pore Size: 3UM
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN DURASHELL C18 ANAL 3UM 3.0X100MM 75807-068 DC931003-0 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty