Would you like to log in again?
COLUMN XBPC18-L 3UM 4.6 X 10MM FOR HPLC
Supplier: Agela Technologies
Venusil* XBP C18(L) Column 4.6x10mm^ Venusil* XBP C18(L) Column, Type: Analytical, For HPLC, Has relatively low surface area which could shorten the retention time and analysis time, Surface Area: 200 m2/g, Pore Size: 150 A, Particle Size: 3um, Dimension: 4.6x10mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN XBPC18-L 3UM 4.6 X 10MM FOR HPLC 75807-778 VX950105-LS Pack of 4 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty