Would you like to log in again?
COLUMN FLASH SPHERICAL HILIC 20UM 120G
Supplier: Agela Technologies
Flash Column, SH, Spherical HILIC, Particle Size: 20um, Pore Size: 100A, Sorbent Size: 120g^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN FLASH SPHERICAL HILIC 20UM 120G 75807-890 SH120120-0 Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty