Would you like to log in again?
COLUMN FLASH C18 SO-2 20-35UM 100A 330G
Supplier: Agela Technologies
Claricep* Spherical Silica SO-2 C18 flash column 330g^ Claricep* Spherical Silica, SO-2, C18 flash column, Hand screw-on head cap, Higher pressure tolerance (180 psi), Choice of solid sample loading, Conventional or Luer lock fittings Average Particle, Size: 20-35 um, Pore Size: 100 A, Size: 330g
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN FLASH C18 SO-2 20-35UM 100A 330G 75807-956 SO230330-2 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty