Would you like to log in again?
MICROPLATE PPT 96 WELL CLEANERT 2 ML
Supplier: Agela Technologies
Cleanert* Microplate PPT Protein Precipitation Plate 2 ML^ Cleanert* Microplate PPT Protein Precipitation Plate, Features: Rapid and high throughput protein precipitation for bio-samples such as plasma, serum, 96-well parallel processing and automation, Suitable for both acetonitrile, Size: 2 ML
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MICROPLATE PPT 96 WELL CLEANERT 2 ML 75808-488 96CD2025-Q Pack of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty