Would you like to log in again?
COLUMN SPHCL C18 CLARICEP FLSH 120G 20UM
Supplier: Agela Technologies
Claricep* Spherical Silica C18 flash column 120g^ Claricep* Spherical Silica, C18 flash column, Hand screw-on head cap, Higher pressure tolerance (180 psi), Choice of solid sample loading, Conventional or Luer lock fittings Average Particle, Size: 20 um, Pore Size: 100 A, Size: 120g
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN SPHCL C18 CLARICEP FLSH 120G 20UM 75808-600 SS120120-0 Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty