Would you like to log in again?
COLUMN INNOVAL SILICA 10X10MM 5 UM 100 A
Supplier: Agela Technologies
Column Innoval Silica 10X10Mm 5umm 100 A^ Column Innoval Silica, Dimension: 10X10Mm, Particle size: 5um 100 A, packed with ultra pure silica particles which are prepared by an aggregation of ultra pure silica sols, shows excellent reproducibility and high efficiency, Metal Impurity<30ppm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN INNOVAL SILICA 10X10MM 5 UM 100 A 75808-814 ISI950110-0S Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty