Would you like to log in again?
COLUMN UNISOL C18 (2) 3 UM 100A 2.1X50MM
Supplier: Agela Technologies
Unisol C18(2) Column 2.1x50mm^ Unisol C18(2) Column, Improved version of Unisol C18 phase, Excellent balance of hydrophobicity, retention, resolution, contamination tolerance, provides reproducibility, Low bleeding, High sensitivity for LC-MS, Particle Size: 3um, Pore Size: 100A, Dimension: 2.1x50mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN UNISOL C18 (2) 3 UM 100A 2.1X50MM 75808-856 UO930502-2 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty