Would you like to log in again?
Surgipath® Alcian Blue Stain kit, Leica
Supplier: Leica
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Surgipath®
38016SS3 38016SS5
75810-404KT 542.84 USD InStock
75810-404 75809-288
Surgipath® Alcian Blue Stain kit, Leica
Stains Histology Stains
Combines the components of the Alcian Blue and Periodic Acid Schiff (PAS) stain kits.
  • pH 2.5
  • For identifying acids
Alcian Blue is normally prepared at an acidic pH of 2.5 and is used to identify acid mucopolysaccharides and acetic mucins.
Excessive amounts of non-sulfated acidic mucosubstances are seen in mesotheliomas, certain amounts occur normally in blood vessel walls but increase in early lesions of atherosclerosis.
Ordering information: The Alcian Blue Stain Kit contains 500 mL each of Periodic Acid, Schiffs Reagent, and Alcian Blue. The Alcian Blue Stain Kit (Special) contains 500 mL each of Alcian Blue and Nuclear Fast Red.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty