Would you like to log in again?
Surgipath® Biopsy Cassettes, Leica
Supplier: Leica
Surgipath®
3802469 3802447 3802467 3802449 3802451
75809-308CS 195.58 USD InStock
75809-308 75809-310 75809-302 75809-304 75809-306
Surgipath® Biopsy Cassettes, Leica
Embedding Cassettes Biopsy Processing Cassettes
Biopsy Cassettes are designed to allow processing of the smallest specimens without the need to use foam inserts or lens paper.
  • Available in 5 colors
  • 1 x 1 mm square holes
  • 50° writing surface
  • Attached, hinged two piece design
The rigidly constructed cassettes are made of solvent and acid resistant plastic featuring a textured writing surface on three sides.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty