Would you like to log in again?
Surgipath® ActivFlo Biopsy II Cassettes, Leica
Supplier: Leica
Surgipath®
39LC-610-4-L 39LC-610-2-L 39LC-610-9-L 39LC-610-6-L 39LC-610-5-L
75809-470CS 440.93 USD InStock
75809-470 75809-472 75809-464 75809-466 75809-468
Surgipath® ActivFlo Biopsy II Cassettes, Leica
Embedding Cassettes Biopsy Processing Cassettes
ActivFlo Biopsy II cassettes feature a patented “Fluidic Flow” pore design for maximum reagent exchange.
  • Available in 5 colors
  • Validated for Leica IP C Cassette Printer
  • <1% printing error rate
  • Two-piece, lids packaged separately
  • 45° writing surface
With a 0.67 mm pore size, the cassettes include an innovative V-shape pore design that pulls fluid in and forces air out. The cassettes feature sloped walls, creating a basket for small tissue management.
Packaging: Lids packaged separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty