Would you like to log in again?
Surgipath® IP ActiveFlo Biopsy III Cassettes, Leica
Supplier: Leica
Surgipath®
39LC-625-7-L 39LC-625-2-L 39LC-625-8-L 39LC-625-10-L 39LC-625-11-L 39LC-625-5-L 39LC-625-4-L 39LC-625-3-L 39LC-625-1-L 39LC-625-9-L 39LC-625-6-L
75806-620CS 318.97 USD InStock
75806-620 75806-608 75806-618 75809-474 75809-476 75806-614 75806-606 75806-616 75806-622 75806-610 75806-612
Surgipath® IP ActiveFlo Biopsy III Cassettes, Leica
Embedding Cassettes Biopsy Processing Cassettes
IP ActivFlo Biopsy III cassettes feature a patented “Fluidic Flow" pore design for maximum reagent exchange.
  • Available in 11 colors
  • Validated for Leica IP C Cassette Printer
  • <1% printing error rate
  • Two piece cassettes with hinged lid
With a 0.26 mm pore size, the cassettes can secure the smallest specimens, eliminating the need for biopsy bags and sponges.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty