Would you like to log in again?
Surgipath® Double-Frost Beveled-Edge Microscope Slides, Leica
Supplier: Leica
Surgipath®
3800375
75809-506CS 354.57 USD InStock
75809-506
Surgipath® Double-Frost Beveled-Edge Microscope Slides, Leica
Slides Microscope Slides
Double frosted Beveled Edge Slides feature beveled edges and corners that make for easier handling.
  • Beveled edges
  • Easy handling
  • For blood smears
Slides are good for blood smears, producing a feathered edge. Double frosted surface allows writing on either side of the slide. Each slide is washed and carefully inspected to ensure quality.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty