Would you like to log in again?
Surgipath® Snowcoat® Clipped Corner X-Tra® Microscope Slides, Leica
Supplier: Leica
Surgipath®
3800062CL 3800061CL 3800063CL 3800060CL 3800056CL
75809-538CS 559.99 USD InStock
75809-538 75809-530 75809-532 75809-534 75809-536
Surgipath® Snowcoat® Clipped Corner X-Tra® Microscope Slides, Leica
Slides Microscope Slides
Snowcoat® Clipped Corner X-Tra Slides are clipped at 45° angles to help reduce glass breakage and fragmentation when loaded into slide printers.
  • Available in 5 colors
  • Positive charged adhesion
  • Optimized for Leica IP-S Printers
These slides have specialty adhesion for Histology application.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty