Would you like to log in again?
COLUMN UNISOL C18 (2) 5UM 4.6X150MM
Supplier: Agela Technologies
Unisol C18(2) Column 4.6x150mm^ Unisol C18(2) Column, Improved version of Unisol C18 phase, Excellent balance of hydrophobicity, retention, resolution, contamination tolerance and provides excellent reproducibility, Low bleeding, High sensitivity for LC-MS, Particle Size: 5um, Dimension: 4.6x150mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN UNISOL C18 (2) 5UM 4.6X150MM 75809-882 UO951505-2 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty