Would you like to log in again?
COLUMN C18 5UM 4X250MM
Supplier: Agela Technologies
Unisol C18 Column 4x250mm^ Unisol C18 Column, A Slightly Polar, 100% Water Compatible and Universal Reversed Phase, Robust and reproducible performance, Wide pH range, Low bleed and High sensitivity for LC-MS, Great inertness and efficiency, Particle Size: 5um, Pore Size: 100A, Dimension: 4x250mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN C18 5UM 4X250MM 75809-886 UO952504-0 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty