Would you like to log in again?
Surgipath® Prefilled Formalin Specimen Jars, Leica
Supplier: Leica
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Surgipath®
3800780 3800810 3800759 380075432AS 380077027 380081018 3800840 3800790
75810-270CS 102.45 USD InStock
75810-270 75810-268 75810-266 75810-298 75810-276 75810-296 75810-272 75810-294
Surgipath® Prefilled Formalin Specimen Jars, Leica
Jars Specimen Jars
Pre-filled Formalin Specimen Jars are impact-resistant, clear polypropylene jars prefilled with 10% Neutral, Phosphate Buffered Formalin.
  • Wide mouth
  • Ribbed caps
  • Leakproof
  • Great for small specimens
  • Easy to store and ship
Wide-mouth jars provide easy access to specimens
AffirmSeal jars are made of durable plastic to prevent cracking when inadvertently dropped, and comply with 95 kPa pressure testing for safe air shipment and use in pneumatic transport systems.
The amount of formalin in the specimen jars is approximately half the volume of the jar. Caps are ribbed, leakproof and easy to open or close even with wet hands or gloves. Lid provides an audible and tactile acknowledgement when jar is closed properly.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty