Would you like to log in again?
Surgipath® Fix-All Tissue Fixative, Leica
Supplier: Leica
Surgipath®
3800686
75810-338CS 232.71 USD InStock
75810-338
Surgipath® Fix-All Tissue Fixative, Leica
Stains Microscopy and staining reagents
Fix-All is a tissue fixative containing alcohol, barium chloride and 10% formalin.
  • Recommended for all tissues
  • Tinted light yellow
  • Substitute for B-5
It is recommended for fixation of all types of tissues, including biopsies, and is particularly good for fixing dense and fatty tissues. Routine, special and immunohistochemical procedures yield excellent results. Fix-All is tinted a light yellow (color) for differentiation. Fix-All can be readily substituted for B-5 fixative.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty