Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin Y (yellowish), powder, Surgipath®, certified by the Biological Stain Commission

Supplier: Leica Biosystems
Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

Surgipath®
3803805
75810-364EA 119.13 USD InStock
75810-364
Eosin Y (yellowish), powder, Surgipath®, certified by the Biological Stain Commission
Eosin Y (yellowish)
Powdered Stain Eosin Y Dye can be used to prepare Eosin counterstains for Romanovsky techniques and for making EA formulations.

  • Alcohol and water soluble
  • Certified by the Biological Stain Commission
  • Colour Index Number 45380

The total dye content is stated on each label.

Certifications: Dye is certified by the Biological Stain Commission.


Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
MDL Number: MFCD00005040
CAS Number: 17372-87-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty