Would you like to log in again?
Surgipath® Modified Perls Prussian Blue Iron Stain Kit, Leica
Supplier: Leica
Surgipath®
38016SS7
75810-406KT 586.94 USD InStock
75810-406
Surgipath® Modified Perls Prussian Blue Iron Stain Kit, Leica
Stains Histology Stains
Perls’ Prussian Blue Iron Kit stains are used to detect and identify ferric (Fe3+) iron in tissue preparations, blood smears, or bone marrow smears.
  • To detect ferric iron
  • For use with tissues, blood, or bone marrow
Minute amounts of ferric acid are commonly found in bone marrow and in the spleen.
Ordering information: This kit contains 500 mL each of Potassium Ferrocyanide Solution, Hydrochloric Acid Solution, and Nuclear Fast Red.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty