Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 37% NF

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Muriatic acidHydrochloric acid solution

HY104-2.5LTPV HY104-20LT HY104-500ML-CS HY104-2.5LTGL HY104-2.5LTG-C HY104-4LTPV-CS HY104-200LTB HY104-500MLGL HY104-2.5LPV-C HY104-4LTPV
700000-568EA 1822.74 USD InStock
700000-568 75811-378 75811-368 75810-486 75811-374 700000-570 75811-376 75811-380 75811-370 75811-372
Hydrochloric acid 37% NF
Hydrochloric acid
Hydrochloric Acid, 37 Percent, NF is an excellent acidifying reagent. 

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 


Formula: HCl
MW: 36.46 g/mol
Density: 1.200
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay (w/w) 36.5 - 38.0 %
Assay (w/w) 36.5 - 38.0 %
Bromide or Iodide To Pass Test
Bromide or Iodide To Pass Test
Free Bromine or Chlorine To Pass Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty