Synonyms:
Muriatic acid
HY106-4LT
HY106-2.5LTG
HY106-4LT-CS4
HY106-20LT
HY106-2.5CS4
HY106-200LTBL
HY106-500MLGL
HY106-500ML-CS
95036-672CS
585
USD
InStock
95036-672
75811-390
75811-388
95036-674
75811-384
75811-386
75811-392
75811-382
Hydrochloric acid 37% FCC
Hydrochloric acid
Hydrochloric Acid, 37 Percent, FCC is used in food processing, or as a food additive to adjust the pH. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Corrosion;Exclamation mark
UN1789