Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Dichloromethane ≥99.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Methane dichlorideMethylene chloride

M1172-100ML M1172-1LT M1172-500CS6 M1172-500MLGL M1172-4LT M1172-200LTBL M1172-4CS4 M1172-20LT M1172-1CS6
95031-330EA 672 USD InStock
95031-330 95030-908 75811-598 95030-906 95030-904 75811-594 95030-910 95031-328 75811-596
Dichloromethane ≥99.0% FCC
Dichloromethane
Methylene Chloride, FCCis used to decaffeinate coffee and tea as well as to prepare extracts of hops and other flavorings. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

GHS hazard : Exclamation mark;Health hazard
UN1593


Formula: CH₂Cl₂
MW: 84.93 g/mol
Density: 1.326
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
UN: 1593
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg
Order Now
Specification Test Results
Assay 99.0 %
Distilling Range 39.0 - 41.0 °C
Specific Gravity 1.318 -1.323
Acidity (As HCl) max. 10 mg/kg
Free Halogens To Pass Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty