Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tri-Sodium citrate dihydrate ≥99.0%, granulated FCC

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Sodium Citrate, DihydrateCitric acid trisodium salt dihydrateSodium Citrate DihydrateSodium citrate tribasic dihydrateTrisodium citrateSodium citrate dihydrateCitric acid tridosium salt dihydrate

S1256-2.5KG S1256-45KGBL S1256-500CS6 S1256-2.5CS4 S1256-12KG S1256-500GM
95035-796EA 151 USD InStock
95035-796 75812-212 95035-792 95035-794 95035-790 75812-210
tri-Sodium citrate dihydrate ≥99.0%, granulated FCC
tri-Sodium citrate dihydrate
Sodium Citrate, Dihydrate, Granular, FCC is used as a flavorant and preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/03/16640603.jpg
Order Now
Specification Test Results
Assay (Anhydrous Basis) 99.0 - 100.5 %
Water 10.0 - 13.0 %
Alkalinity To Pass Test
Lead (Pb) max. 2 mg/kg
Identification To Pass Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty