Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ≥95.0%, pellets FCC

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

S1303-2.5KG-CS S1303-500GM-CS S1303-12KG S1303-2.5KG S1303-50KGBL S1303-500GM
75812-236EA 412.08 USD InStock
75812-236 75812-244 75812-234 75812-238 75812-242 75812-240
Sodium hydroxide ≥95.0%, pellets FCC
Sodium hydroxide
Sodium Hydroxide, Pellets, FCC is used in the chemical washing and peeling of fruits and vegetables and as a thickening agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

GHS hazard : Corrosion;Exclamation mark
UN1823


Formula: NaOH
MW: 40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Assay (Total Alkali As NaOH) 95.0 - 100.5 %
Arsenic (As) max. 3 mg/kg
Carbonate (As Na₂CO₃) max. 3.0 %
Insoluble Substances & Organic Matter To Pass Test
Lead (Pb) max. 2 mg/kg

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty