Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
S1303-2.5KG-CS
S1303-500GM-CS
S1303-12KG
S1303-2.5KG
S1303-50KGBL
S1303-500GM
75812-236EA
404
USD
InStock
75812-236
75812-244
75812-234
75812-238
75812-242
75812-240
Sodium hydroxide ≥95.0%, pellets FCC
Sodium hydroxide
Sodium Hydroxide, Pellets, FCC is used in the chemical washing and peeling of fruits and vegetables and as a thickening agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Corrosion;Exclamation mark
UN1823