Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tri-Sodium citrate dihydrate ≥99.0%, granulated USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Sodium Citrate, DihydrateCitric acid trisodium salt dihydrateSodium Citrate DihydrateSodium citrate tribasic dihydrateTrisodium citrateSodium citrate dihydrateCitric acid tridosium salt dihydrate

SO165-50KGBL SO165-12KG SO165-500CS6 SO165-2.5KG SO165-12KG SO165-500GM SO165-2.5CS4
95035-798CS 1708.5 USD InStock
95035-798 75812-436 95035-804 89050-470 95035-800 CA89050-470 95035-802
tri-Sodium citrate dihydrate ≥99.0%, granulated USP
tri-Sodium citrate dihydrate
Sodium Citrate, Dihydrate, Granular, USP can be used as an alkalizing agent, buffering agent, emulsifier, or sequestering agent. 

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/03/16640603.jpg
Order Now
Specification Test Results
Assay (C₆H₅Na₃O₇) 99.0 - 100.5 %
Water 10.0 - 13.0 %
Alkalinity To Pass Test
Tartrate To Pass Test
Heavy Metals max. 0.001 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty