Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ≥95.0%, pellets NF

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

SO170-2.5KG-CS SO170-12KG SO170-500GM-CS SO170-50KGBL SO170-500GM SO170-2.5KG
89050-478EA 202.98 USD InStock
89050-478 75812-448 75812-446 BAL89050476KPH 89050-482 75812-450
Sodium hydroxide ≥95.0%, pellets NF
Sodium hydroxide
Sodium Hydroxide, Pellets, NF is used as a pH adjusting agent. 

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 


Formula: NaOH
MW: 40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Assay (Total Alkali Including A 95.0 - 100.5 %
Insoluble Substances & Organic Matter To Pass Test
Potassium (K) To Pass Test
Heavy Metals max. 30 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty