Glass pressure tubes are rated at 150psig at 120°C
The tubes feature a #7 Ace-Thred sample port, and offer the convenience of sampling while remaining connected to the researcher's apparatus. Observe all laboratory safety procedures, including working behind a shield, as glass vessels are not guaranteed from breakage due to varying conditions.
Ordering information: Tubes are available with either no plugs, front-seal plugs, or back-seal plugs.