Would you like to log in again?
Gas-Tight Stainless Steel Drum Pumps, Burkle
Supplier: Bürkle
5611-1090 5611-0090
75813-134EA 847.31 USD InStock
75813-134 75813-136
Gas-Tight Stainless Steel Drum Pumps, Burkle
Pumps Drum/transfer Pumps
Gas-tight barrel pump with flexible discharge hose for filling off into large canisters and containers.
  • For bottle openings from 16 - 49 mm
  • Immersion tube Ø 32 mm
  • Gas-tight barrel screw joints with 2" fine thread
  • The pump can be hermetically sealed with two ball valves
Convenient transfer because the canister can stay on the floor. The connection cone is held onto the container. Parts in contact with media - stainless steel (1.4301), PTFE, nickel-plated brass, PA that can conduct electricity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty