Would you like to log in again?
Pressure Tubes with #7 Ace-Thred Sample Port, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8649-10 8649-112 8649-130 8649-217 8649-146 8649-120 8649-24 8649-233 8649-239 8649-137 8649-224 8649-230 8649-28 8649-22 8649-15 8649-215 8649-145 8649-35 8649-40 8649-122 8649-30 8649-12 8649-232 8649-37 8649-222 8649-245 8649-117 8649-139 8649-14 8649-210 8649-114 8649-212 8649-115 8649-128 8649-235 8649-214 8649-135 8649-132 8649-240 8649-246 8649-32 8649-46 8649-110 8649-33 8649-26 8649-133 8649-20 8649-124 8649-226 8649-126 8649-237 8649-39 8649-140 8649-45 8649-220 8649-228 8649-17
75813-250EA 272.19 USD InStock
75813-250 75813-130 75813-252 75813-132 75813-122 75813-200 75813-244 75813-124 75813-246 75813-202 75813-126 75813-204 75813-248 75813-128 75813-206 75813-208 75813-240 75813-120 75813-242 75813-232 75813-234 75813-236 75813-116 75813-238 75813-118 75877-648 76318-592 76318-594 75813-190 75813-192 75813-194 75813-196 75813-230 75813-198 75813-188 75813-222 75813-224 75813-226 75813-228 75813-180 75813-182 75813-260 75813-184 75813-262 75813-186 75813-220 75813-264 75813-254 75813-210 75813-178 75813-256 75813-212 75813-214 75813-258 75813-216 75813-218 76318-596
Pressure Tubes with #7 Ace-Thred Sample Port, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
Glass pressure tubes are rated at 150psig at 120°C
The tubes feature a #7 Ace-Thred sample port, and offer the convenience of sampling while remaining connected to the researcher's apparatus. Observe all laboratory safety procedures, including working behind a shield, as glass vessels are not guaranteed from breakage due to varying conditions.
Ordering information: Tubes are available with either no plugs, front-seal plugs, or back-seal plugs.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty