Would you like to log in again?
EnviroTrans™ Swab Rinse Kit, Barney Miller Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
SRK100
75813-998PK 73.83 USD InStock
75813-998
EnviroTrans™ Swab Rinse Kit, Barney Miller Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
EnviroTrans™ Swab Rinse Kit, Barney Miller Broth is recommended for environmental monitoring.
  • Collect and transport environmental samples with efficiency and ease.
  • Prefilled with Barney Miller Broth
Recommended for environmental monitoring.
Collect and transport environmental samples with efficiency and ease. Simply remove the cap and attached Dacron® swab, collect the sample, recap, and send to the lab. Each EnviroTrans™ tube contains broth or buffer.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty