Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

MLA Macro Volume Pipettors, VistaLab

Supplier: VistaLab
1079 1066 1061 1077 1078 1069 1134 1070 1076 1067 1068 1065
53514-058EA 528.13 USD InStock
53514-058 75814-394 53514-048 75813-800 53514-050 53514-052 75814-400 75814-398 53514-054 53514-078 75814-396 53514-056
MLA Macro Volume Pipettors, VistaLab
Pipettes Repetitive Pipettors
MLA macro volume pipettors offer larger pipetting volumes than traditional models, with single-stroke 'to deliver' operation.

  • Available in a range of fixed and adjustable volumes
  • Different colors available for customizing models

MLA pipettors are made of precision-engineered aluminum alloy and stainless steel

Adjustable model available with a range from 1 to 5 ml in 0.1-ml increments.

Selectable model available with a range of set volumes at 2, 3, 4, and 5 ml.

MLA macro volume pipettors are available in 10 fixed volumes ranging from 1 to 10 ml.

Accessories information: Seal Kits for MLA Pipettors are available separately. Seal kits contain lubricant, seals, and instructions.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty