Would you like to log in again?
GLOVE G-TEK 13GUG SEAMLESS KNIT GY LARGE
Supplier: PIP
Glove G-TEK* Large^ Glove, G-Tek* CR, 13 Gauge, Material: HPPE/glass shell with polyurethane coated palm and fingers, Seamless Knit, Gray, Construction: Seamless Knit, Applications: Electronics, Glass cutting, Sheet metal handling, General duty, Parts assembly, Hem Color: Brown, Size: Large
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVE G-TEK 13GUG SEAMLESS KNIT GY LARGE 75814-406 16-560/L Case of 72 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty