Would you like to log in again?
Phenol Red Broth with Trehalose, Hardy Diagnostics
Supplier: Hardy Diagnostics
Y317
75814-676PK 1179.65 USD InStock
75814-676
Phenol Red Broth with Trehalose, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Phenol Red Broth with Trehalose is used for the differentiation of microorganisms by carbohydrate fermentation.
  • Contains a durham tube for gas detection
  • Differentiation of microorganisms by carbohydrate fermentation
For the differentiation of microorganisms by carbohydrate fermentation. Contains a durham tube for gas detection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty