Would you like to log in again?
Tech Agar Tube Slant, Hardy Diagnostics
Supplier: Hardy Diagnostics
L96
75814-680PK 196.44 USD InStock
75814-680
Tech Agar Tube Slant, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Tech Agar Tube Slant is used for the production of pyocyanin pigment by Pseudomonas species.
  • Pyocyanin pigment production by Pseudomonas species
Ordering information: Minimum order of 5 packs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty