Would you like to log in again?
Mueller Hinton Agar with Blood, Hardy Diagnostics
Supplier: Hardy Diagnostics
H21
75814-684PK 85.07 USD InStock
75814-684
Mueller Hinton Agar with Blood, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Mueller Hinton Agar with Blood is used for disk diffusion sensitivity testing of Streptococcus spp.
  • Disk diffusion sensitivity training
  • With 5% sheep blood
Mueller Hinton Agar with 5% Sheep Blood For disk diffusion sensitivity testing of Streptococcus spp.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty