Would you like to log in again?
WST-1 Cell Proliferation Assay Kit, Cayman Chemical
Supplier: Cayman Chemical
10008883-1 10008883-5
75816-716EA 166.24 USD InStock
75816-716 75816-714
WST-1 Cell Proliferation Assay Kit, Cayman Chemical
Assays Cellular Assays
Used for detection of viable cells.
  • A colorimetric, non-radioactive assay for assessing cell viability and proliferation
  • Tetrazolium salt, WST-1, is reduced to a soluble purple formazan by trans-plasma membrane electron transport from NADH via an electron mediator
  • Detect 1000 to 200000 cells per well
  • Rapid assay; get results in 2 to 4 hours
Cayman’s WST-1 Proliferation Assay is based on the reduction of tetrazolium salt WST-1 to a soluble formazan dye by electron transport across the plasma membrane of dividing cells. This kit will also allow investigators to screen drug candidates involved in cell cycle regulation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty